missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dodecyl Sulfate, MP Biomedicals™

Catalog Number 11467140
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium Dodecyl Sulfate, 20% solution, Liquid

  • SDS 20% solution, ready to use solution is a multipurpose, anionic detergent used for protien solubilization,plasmid extraction from bacteria and to reduce non-specific binding during nucleic acid hybridization on membranes.
  • SDS is an anionic detergent typically used to solubilize and denature proteins for electrophoresis.
  • Most proteins bind SDS in a ratio of 1.4 grams SDS to 1 gram protein.
  • The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS.
  • It is RNAse,DNAse, Protease free with an assay value of 20%.
  • This solution is provided as a convenient ready-to-use format which eliminates the need to handle powdered SDS.
CAS 151-21-3
Physical Form Liquid
Molecular Formula C12H25NaO4S
Concentration 20% Solution
MDL Number MFCD00036175
Quantity 1 L
Beilstein 3599286
Synonym SDS Solution
Solubility Information Soluble in water.
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'