missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Carbonate, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 500g, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M

Brand:  Fisher Chemical S/4240/53

Code : 22

Additional Details : CAS Number : 144-55-8 Weight : 0.50000kg

Packaging Plastic powder jar
Quantity 500g

 View more versions of this product

Product Code. 10020510

Quantity Price
1 25.36 € / 500g
4 22.21 € / 500g
16 21.09 € / 500g
In stock, ready to ship: 38

Please to purchase this item. Need a web account? Register with us today!



Description

Description

CHNaO3, CAS Number-144-55-8, acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogencarbonate, sodium hydrogen carbonate, 500g, CHEBI:32139, White, 84.01g/mol, UIIMBOGNXHQVGW-UHFFFAOYSA-M

Specifications

Chemical Identifiers

144-55-8
84.006
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium;hydrogen carbonate
CHNaO3
3528
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
CHEBI:32139
C(=O)(O)[O-].[Na+]

Specifications

Powder Solid
CHNaO3
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
C(=O)(O)[O-].[Na+]
84.006
CHEBI:32139
Plastic powder jar
270°C
500g
144-55-8
3528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
84.01g/mol
White
8.3
Videos
SDS Documents
Certificates
View accessory products