missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Carbonate, Extra Pure, SLR, Fisher Chemical™
13.69 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
1 kg
3 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
Sodium Hydrogen Carbonate, Extra Pure, SLR
Specifications
Melting Point | 270°C |
Color | White |
pH | 8.3 |
Physical Form | Powder/Solid |
Quantity | 1 kg |
IUPAC Name | sodium;hydrogen carbonate |
Formula Weight | 84.01g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'