missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sodium iodide, ultra dry, 99.999% (metals basis)

Brand:  Alfa Aesar™ 013604.22

Code : NEW

Additional Details : CAS Number : 7681-82-5 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15476015

  • 494.00 € / 100g

Estimated Shipment date
from Supplier 17-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Ampouled under argon
Specifications

Chemical Identifiers

7681-82-5
149.894
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
5238
sodium;iodide
INa
MFCD00003532
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
CHEBI:33167
[Na+].[I-]

Specifications

Sodium iodide
7681-82-5
INa
14,8631
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium;iodide
5238
149.89
99.999%
−10 Mesh
Ampule
Light Sensitive & Hygroscopic
White
100g
Ultra dry
(metals basis)
MFCD00003532
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
[Na+].[I-]
149.894
CHEBI:33167
Beads
3.67g/mL
Odorless
1.7745
1300°C to 1304°C
651°C
Videos
SDS Documents