missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium methoxide, ACS reagent, 0.5M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
51.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 65.0°C |
Melting Point | -98.0°C |
Quantity | 100 mL |
Linear Formula | CH3ONa |
Flash Point | 11°C |
Merck Index | 15,8775 |
Solubility Information | Solubility in water: reacts |
Formula Weight | 54.02 |
Grade | ACS Reagent |
Packaging | AcroSeal™ Glass bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'