missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium nitrite, 99%, extra pure, contains an anticaking reagent, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Physical Form | Crystalline Powder |
Quantity | 2.5 kg |
Assay Percent Range | 99% |
Linear Formula | NaNO2 |
Fieser | 01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86 |
Merck Index | 15, 8779 |
Formula Weight | 69 |
Percent Purity | ≥98.5% |
Grade | Extra Pure |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'