missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitroprusside Dihydrate, puriss. p.a., ≥99%, ACS reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 g |
Linear Formula | Na2[Fe(CN)5NO] · 2H2O |
UN Number | UN3288 |
Formula Weight | 297.95g/mol |
Percent Purity | ≥99% |
Chemical Name or Material | Sodium nitroprusside dihydrate |
Safety and Handling
Product Identifier
- Sodium nitroprusside dihydrate
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'