missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Peroxide, Reagent grade, Honeywell™
Click to view available options
Quantity:
100 g
500 g
Description
- ≥95% for general laboratory use.
Specifications
Physical Form | Granules |
Quantity | 100 g |
UN Number | 1504 |
Formula Weight | 77.98g/mol |
Percent Purity | 97% |
Grade | Reagent |
Chemical Name or Material | Sodium Peroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'