missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, Anhydrous, Powder, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- For Liquid Chromatography and Molecular Biology applications
Specifications
Physical Form | Powder |
Quantity | 2.5 kg |
Formula Weight | 141.96 |
Grade | Ultrapure Bioreagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.07kg |
Chemical Name or Material | Sodium Phosphate, Dibasic, Anhydrous |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'