missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate Anhydrous, 99+%, Extra Pure, Fisher Chemical™
Click to view available options
Quantity:
500 g
3 kg
5 kg
Description
Sodium Sulfate Anhydrous, 99+%, Extra Pure
Specifications
Melting Point | 884°C |
Color | White |
pH | 5 to 8 |
Physical Form | Solid |
Quantity | 500 g |
IUPAC Name | disodium;sulfate |
Formula Weight | 142.04g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'