missing translation for 'onlineSavingsMsg'
Learn More

Sodium Sulfate Anhydrous, min.99%, 12-60 Mesh, Granular, Ultra Resi-Analyzed, ACS Reagent, J.T. Baker™

Catalog Number p-4665838
Click to view available options
Quantity:
500 g
2.5 kg
45.5 kg
Packaging:
Glass Bottle
Lined Fiber Drum
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7757-82-6
Na2O4S
142.04
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
24436
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
pH 5.2 to 9.2
Physical Form Granules
Quantity 500 g
IUPAC Name disodium;sulfate
Formula Weight 142.04
Grade Meets ACS reagent chemical requirements
Loss on Ignition max. 0.5%
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.68kg
Chemical Name or Material Sodium Sulfate, Anhydrous
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'