missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-pentoxide, 95%, pure, Thermo Scientific Chemicals

Catalog Number p-192759
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
14593-46-5
C5H11NaO
110.13
MFCD00064245
CGRKYEALWSRNJS-UHFFFAOYSA-N
sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
2733478
sodium;2-methylbutan-2-olate
CCC(C)(C)[O-].[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
14593-46-5
C5H11NaO
110.13
MFCD00064245
CGRKYEALWSRNJS-UHFFFAOYSA-N
sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
2733478
sodium;2-methylbutan-2-olate
CCC(C)(C)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Cream or Tan
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3CH2C(CH3)2ONa
Quantity 100 g
Formula Weight 110.13
Percent Purity 95%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium tert-pentoxide, 95%
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium tert-pentoxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H251-Self-heating: may catch fire.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235+P410-Keep cool. Protect from sunlight.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'