Learn More
Sodium thiosulfate, 0.1 N standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 124270025
44.86 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : 29
Additional Details : CAS Number : 7772-98-7 Weight : 2.55000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent, Volumetric standard solutionSpecifications
Sodium thiosulfate | |
7772-98-7,7732-18-5 | |
2.5 L | |
Na2S2O3 | |
15, 8821 | |
Solubility in water: soluble in water. Other solubilities: practically insoluble in alcohol | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
158.11 | |
Plastic Bottle |
Colorless | |
Liquid | |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium sulfanidesulfonate | |
24477 | |
0.0950 to 0.1050N (20°C) |
RUO – Research Use Only