missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 98.5%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4509852
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
7772-98-7
Na2O3S2
158.10
MFCD00003499
AKHNMLFCWUSKQB-UHFFFAOYSA-L
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
24477
[Na+].[Na+].[O-]S([S-])(=O)=O
7772-98-7
Na2O3S2
158.10
MFCD00003499
AKHNMLFCWUSKQB-UHFFFAOYSA-L
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
24477
[Na+].[Na+].[O-]S([S-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 48.0°C
Color White
pH 6.0 to 8.5 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 98% min. (on dry substance) (Iodimetry)
Linear Formula Na2S2O3
Merck Index 15,8821
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Formula Weight 158.1
Percent Purity 98.5%
Grade Extra Pure
Loss on Drying 2% (105°C, 2 hrs) max.
Packaging Plastic bottle
Chemical Name or Material Sodium thiosulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'