Learn More
Sodium Thiosulfate Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
H10Na2O8S2, CAS Number-10102-17-7, ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m, 1L, CHEBI:32150
Brand: Fisher Chemical J/7950/15
38.26 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 23
Additional Details : CAS Number : 10102-17-7 Weight : 1.12000kg
Description
H10Na2O8S2, CAS Number-10102-17-7, ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m, 1L, CHEBI:32150
Ready to use; For volumetric analysisSpecifications
10102-17-7 | |
1 L | |
149186 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
HDPE plastic bottle |
Liquid | |
H10Na2O8S2 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.172 | |
CHEBI:32150 |