missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solutions A+ Aloe Latex Gloves

Catalog Number 15173269
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Protects hands from drying, chapping and cracking. Fisherbrand™ Solutions A+ Aloe Latex Gloves combine protection and comfort.

Latex gloves provide a protective barrier and contain soothing Aloe vera to help prevent skin from drying, cracking and chapping. Textured to provide excellent grip. Powder free. EN 374:2003 (AQL 1.5).
  • PPE Category III
  • Disposable
  • Aloe vera coating protects from drying, chapping and cracking
  • Ideal for prolonged use
  • Textured surface provides good grip
  • Latex
Certifications/Compliance ISO 9001 QMS, FDA Biocompatibility Guidance, Passes Viral Penetration Test (ASTM F1671), 510K Approved
Color Natural
Hand Ambidextrous
Material Latex
Powdered Powder Free
Size Medium
Sterility Non-sterile
Thickness (English) 5.5 mil
For Use With (Application) Ideal for workers who have frequent glove changes
Glove Cuff Style Beaded/Rolled
Quantity 100/Pk.
Tensile Strength 18 MPa
Length (Metric) 240 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'