Learn More
METTLER TOLEDO™ Solutions for perfectION™ Combined ISE
Recommended reference solution for replacing electrolyte in ion selective electrodes.
Brand: METTLER TOLEDO™ 51344754
119.07 EUR valid until 2024-05-31
Use promo code "21700" to get your promotional price.
Code : 59
Additional Details : Weight : 0.49000kg
Description
- Keep electrolyte levels high by refilling regularly – when the probe is submerged, the electrolyte level should always be higher than the sample level.
- Routinely refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.
- Reference table above to ensure proper electrolyte is selected for accurate measurement readings.
Specifications
perfectION Electrolyte EIon Electrolyte E | |
perfectION ion electrolyte E, 5 x 60 mL | |
Combined ISE Solution for perfectION | |
perfectION Electrode: Potassium |
Safety and Handling
- X5 perfectION reference filling solution E
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : silver nitrate