Learn More
Cytiva (formerly Pall Lab) SolVac™ Filter Holder
The SolVac™ magnetic filter holder simplifies clean-up and degassing of mobile phase solvents and other solutions.
Brand:  Cytiva (formerly Pall Lab) 4020
Additional Details : Weight : 0.19000kg
Product Code. 17154261
  • 595.00 € / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • For safe and simply mobile phase filtration
  • Ensures analysts can obtain the best possible results while effectively protecting their valuable equipment
Specifications
Specifications
Non-sterile
10.2 cm2
61 cm (2 ft.) feed line tubing, thumb clamp, sinker, vacuum port adapter, 2 membrane seal gaskets, and 2 seal gaskets
47 mm