Learn More
Sortilin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12369-1-AP-20UL
 View more versions of this product
Product Code. 16870054
  • 145.00 € / 20µL
Description
This gene encodes a protein that is a multi-ligand type-1 receptor with similarity to the yeast carboxypeptidase Y sorting receptor Vps10 protein. The encoded protein, a trans-Golgi network (TGN) transmembrane protein, binds a number of unrelated ligands that participate in a wide range of cellular processes, however, it lacks the typical features of a signalling receptor. In the TGN, furin mediates the activation of the mature binding form. The encoded protein consists of a large luminal domain, a single transmembrane segment and short C-terminal cytoplasmic tail. The luminal domain contains a cysteine-rich region similar to two corresponding segments in the yeast Vps10p, the cytoplasmic tail is similar to the corresponding segment of the cation-independent mannose 6-phosphate receptor and the tail also interacts with the VHS domains of GGA (Golgi-associated, gamma-adaptin homologous, ARF-interacting) proteins.
Specifications
Specifications
Sortilin
Polyclonal
Unconjugated
SORT1
100 kDa NT receptor, Glycoprotein 95, Gp95, Neurotensin receptor 3, NT3, NTR3, SORT1, Sortilin, sortilin 1
Rabbit
Antigen Affinity Chromatography
RUO
20661, 6272, 83576
-20°C
Liquid
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54861, Q6PHU5, Q99523
SORT1
Sortilin Fusion Protein Ag2997
20 μL
Primary
Mouse, Human, Rat