missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Mouse, Clone: SOX2/1792, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | SOX2 |
---|---|
Clone | SOX2/1792 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930888
|
EnquireBio
6657-MSM2-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920888
|
EnquireBio
6657-MSM2-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
SOX2 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
6657 | |
Recombinant human SOX2 protein fragment (aa176-305) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
34kDa |
SOX2/1792 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_003097 | |
SOX2 | |
IgG2b κ | |
Protein A/G Purified | |
Nuclear | |
MCF-7 cells or Cervical carcinoma |