missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 (Master Regulator of Epithelial-Mesenchymal Transition) Mouse, Clone: SOX4/2540, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | SOX4 (Master Regulator of Epithelial-Mesenchymal Transition) |
---|---|
Clone | SOX4/2540 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960888
|
EnquireBio
6659-MSM1-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950888
|
EnquireBio
6659-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
SOX4 (Master Regulator of Epithelial-Mesenchymal Transition) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human SOX4 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
47kDa |
SOX4/2540 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
6659 | |
IgG2b κ | |
Protein A/G purified from Bioreactor concentrate. | |
Nuclear | |
Breast or Colon Carcinoma |