Learn More
SP Bel-Art™ SP Bel-Art WYE (Y) Tubing Connecters For 5/16 in. Tubing
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter
Brand:  SP Bel-Art™ 19615-0000
Additional Details : Weight : 0.24300kg
Product Code. 16401314
  • 36.80 € / Pack of 12
In Stock
to see stock.
Add to basket
Description
Description
  • Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Specifications
Specifications
8 mm
39.7 mm
54 mm
Polypropylene
12/Pk.