missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.
86.50 € - 225.00 €
Specifications
Product Type | Bench Protector |
---|---|
Material | Polyethylene |
Absorbancy | 400 mL/m2 |
Width (English) | 20 in. |
Width (Metric) | 508 mm |
Product Code | Brand | Color | Dimensions (L x W) | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (L x W) | Description | Price | Quantity & Availability | |||
11342194
|
Bel-Art™
F24675-1000 |
Orange | 50 ft. x 20 in. (15.2m x 508mm) | One single roll, 50 ft. |
99.20 €
1 roll |
In Stock
Log in to see stock. |
|||
12918837
|
Bel-Art™
F24675-0020 |
White | 24 x 20 in. (610 x 508mm) | Pack of 100 sheets |
225.00 €
Pack of 100 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||
11367234
|
Bel-Art™
F24675-0000 |
White | 50 ft. x 20 in. (15.2m x 508mm) | One single roll, 50 ft. |
86.50 €
Each |
In Stock
Log in to see stock. |
|||
Description
- Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
- Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
- Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
- Roll up and discard in proper waste container after contamination
- Select orange to identify use with biohazard materials
Specifications
Bench Protector | |
400 mL/m2 | |
508 mm | |
Simplify cleanup and safeguard work surfaces from stains, spills and wear |
Polyethylene | |
20 in. | |
Yes |