Learn More
Bel-Art™ SP Scienceware™ Reuseable/Disposable Desiccant Cartridges
Use anywhere dessiccation is needed, including analytical balance cases and precision laboratory instruments. SP Scienceware™ Reuseable/Disposable Desiccant Cartridges fit “Space Saver” desiccators, in addition to other plastic desiccators of similar size.
69.00 € - 109.26 €
Specifications
Product Type Desiccant Cartridge
Material Aluminum
Disposable Yes
For Use With (Application) Use analytical balance cases and precision laboratory instruments
View More Specs
Products 2
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
10576384
missing translation for 'viewDocuments' Promotion Details
Bel-Art™
F42048-0065
6.5 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
11867913
missing translation for 'viewDocuments' Promotion Details
Bel-Art™
F42048-0100
10 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description
Description
  • Inert, noncorrosive and nontoxic silica gel
  • Clear polystyrene dish allows instant visual inspection of silica gel, which changes color from blue to pale pink as it absorbs moisture
  • Highly porous membrane covers the dish, providing excellent moisture absorption
  • Desiccant can be easily regenerated by heating cartridges in a 50°C (122°F) oven overnight
  • For more rapid drying, cut the membrane, transfer the spent gel crystals into a glass or porcelain dish, and place in a 150°C (300°F) oven for one hour
  • Individually foil sealed
  • Fits “Space Saver” dessicator, in addition to other plastic dessicators of similar size
Order Info
Fits Bel-Art Space Saver Desiccator (Mfr. No. 420320000) and other desiccators of similar size.
Specifications
Specifications
Desiccant Cartridge
Aluminum
Yes
Use analytical balance cases and precision laboratory instruments