Learn More
SPE tube silica normal phase polypropylene 500mg, 6mL Bakerbond
Brand:  JT Baker 7086-06
Code : Z3
Additional Details : Weight : 0.19000kg
Product Code. 13174514
  • POA / Pack of 30
Estimated Shipment: 29-05-2024