missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectrum™ Drawer/Ganizer™ Portable Organizer Trays

Rigid 1/8 in. polystyrene trays with easy-gripping folded edges for transporting lab items and mobilizing experimental activities

Brand:  Spectrum™ 155 630

Code : NEW

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15391042

  • 63.00 € / Each


Estimated Shipment
04-02-2019
Add to basket

Description

Description

Deep compartments are ideal for holding large quantities or providing secondary containment for spills. Also designed to fit lab drawers, refrigerators and bench tops to protect supplies, organize valuable space and promote laboratory safety.

  • Designed for portability and storage in drawers
  • Protects supplies, promotes safety and provides secondary containment
  • Six tray configurations available in two sizes (large and small)

Specifications

Specifications

4 Equal 20 x 15cm compartments
0.32cm
Utility and Equipment Organizer Tray
Small
Polystyrene
Videos