missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectrum™ Spectra Mesh™ Woven, Polyester Filters
Consist of individual strands woven into a mesh screen
Brand: Spectrum™ 145 831
Code : NEW
Additional Details : Weight : 0.05000kg
Description
- Individual strands woven into a mesh screen
- Characterized by precise mesh openings, percent open area, and mesh thickness
- Mildly hydrophobic
- Low Adsorption
- Resistant to alklies and organics
- Autoclavable
Specifications
Disc | |
10 μm | |
55 μm |
Polyester | |
Drinking water purification, suspended soils, air/asbestos, coal dust analysis, trace element analysis, fertilizers, aqueous serum clarification, organic serum clarification | |
55 mm |