missing translation for 'onlineSavingsMsg'
Learn More

SPEX CertiPrep™ Method EN15662 SPEXQuE™ QuEChERS Kit

Meet your QuEChERS needs with these general purpose kits

Brand:  SPEX CertiPrep™ EN-CIT-50ML

Code : NEW

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 15266036

  • 277.00 € / 1 unit


Estimated Shipment
07-08-2020
Add to basket

Description

Description

Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.

  • European Methods (EN Method 15662)

Specifications

Specifications

4 g MgSO4,1 g NaCl, 0.5 g sodium citric dibasic sesquihydrate, 1 g sodium citrate tribasic dihydrate
2 mL
EN 15662
Citrate Buffered Extraction Tube
50 Pack
Videos
Documents