missing translation for 'onlineSavingsMsg'
Learn More

SPEX™ SamplePrep 8000 Series Mixer/Mill™ Accessories

For use with 8000D and 8000M Mixer/Mills

Brand:  SPEX™ SamplePrep 8009

Code : NEW

Additional Details : Weight : 5.00000kg

 View more versions of this product

Product Code. 15330522

  • 1840.13 € / 1 set


Estimated Shipment
09-07-2020
Add to basket

Description

Description

Accessories offered:

  • Grinding balls and vials in assorted materials and sizes
  • Grinding ball sets
  • Vial sets with vial and grinding ball(s)
  • Adapters and clamps

Specifications

Specifications

Fe, Cr, Si, Mn, C
Ideal for grinding hard, brittle samples to an analytical fineness, slurry grinding, blending powders and mixing emulsions
7.62cm
7.62cm
Hardened Steel
10g
8000D, 8000M mixer/mills
6.03cm
5.5 to 6
One screw on cap, one O-ring, two 0.5 in. and four 0.25 in. Steel balls
25mL
3 to 10mL
35mL
Grinding Vial Set
Videos
Documents