Learn More
BD Accuri™ Spherotech 8-Peak Validation Beads (FL1 - FL3)
8-Peak validation bead tests are designed to fluoresce in all channels on most flow cytometers and cell sorters, to measure subtle differences in fluorescence levels.
Brand: BD 653144
Additional Details : Weight : 1.00000kg
Description
- Check fluorescence sensitivity and resolution.
Specifications
8-Peak validation beads | |
Validation Beads | |
Flow cytometers and Cell sorters | |
4 mL |