Learn More
Corning™ Spin-X™ Centrifuge Tube Insert Without Membrane
Nonsterile and certified nonpyrogenic
Brand:  Corning™ 9301
Code : A0
Additional Details : Weight : 0.49895kg
Product Code. 10486393
  • 363.82 € / Pack of 1000
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Order Info
Insert only (microfuge tube not included).
Specifications
Specifications
Non-sterile
2 mL