Learn More
Nabertherm™ Split Type Vertical Tube Furnace RSV 1100 Series with B500 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
5920.00 € - 12570.00 €
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17348119
|
Nabertherm™
RI152K2AN |
50 mm | 250 mm |
5920.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17368119
|
Nabertherm™
RI252K2AN |
50 mm | 500 mm |
6420.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17308129
|
Nabertherm™
RI322K2AN |
120 mm | 500 mm |
8000.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17348129
|
Nabertherm™
RI382K2AN |
80 mm | 500 mm |
7760.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17388129
|
Nabertherm™
RI422K2AN |
120 mm | 750 mm |
10120.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17328139
|
Nabertherm™
RI472K2AN |
170 mm | 750 mm |
11330.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17368139
|
Nabertherm™
RI482K2AN |
80 mm | 750 mm |
9670.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17308149
|
Nabertherm™
RI522K2AN |
120 mm | 1000 mm |
10900.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17348149
|
Nabertherm™
RI572K2AN |
170 mm | 1000 mm |
12570.00 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1100°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type N thermocouple
- Frame for vertical operation
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 includes fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- RSV: switchgear and control unit separate from furnace in own wall or standing cabinet
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
B500 | |
Up to 1100°C |