missing translation for 'onlineSavingsMsg'
Learn More

Manutan Stacking chair, Ekwo, blue fabric with black frame

Brand:  Manutan A332603

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12881557

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

A practical stacking conference chair, ideal to use in meetings and training rooms. Easily stacked to save space, stacks in groups of eight.
  • Blue
  • Black frame
  • Seat dimensions (H x W x D): 485mm x 475mm x 415mm
  • Back dimensions (H x W): 310mm x 475mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers