missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMS™ Series 3 Model 400W Cooled Incubator, 450 L, Stainless Steel
Click to view available options
Capacity (Metric):
450 L
Unit Size:
Each
Description
- Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
- Reliable and accurate temperature control
- Standard single and dual temperature, plus programmable models available
- Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
- Self-evaporating condensate drip tray
- White plastic coated wire shelves
- Energy efficient control system and eco-friendly
- P.I.D. heating and cooling to ensure stable internal temperature
- Pre-set safety cutout
- Hermetically sealed refrigeration system
- Door lock
- Digital controls are easy to read and simple to use
- Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
- Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008 |
| Controller Type | Standard digital |
| Description | Freestanding incubator |
| Capacity (Metric) | 450 L |
| Chamber Material | Stainless Steel |
| Convection Type | Fan-assisted Circulation |
| Dimensions (D x W) Shelf | 510 mm x 600 mm |
| Lighting | Optional Fluorescent/UV |
| Material | Stove Enameled Mild Steel |
| No. of Shelves | 3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
