Learn More
Trajan Scientific and Medical™ Stainless-Steel GC Unions
Provide low dead volume connections for optimum chromatography
59.60 € - 62.20 €
Product Code | Brand | Diameter (English) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Outer | Price | Quantity & Availability | |||||
15269103
|
Trajan Scientific and Medical™
103330 |
0.0625 x 0.0625 in. |
62.20 €
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15209103
|
Trajan Scientific and Medical™
101270 |
0.25 x 0.25 in. |
59.60 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
- Low dead volume
- For high temperature applications
- Leak-free connections
- Does not require back ferrules
- Used with Graphite, Graphite Vespel™ or PTFE ferrules
- Suitable for glass, fused silica, metal, plastic or glass lined tubing
- Non-swaging and no tube distortion
Specifications
Stainless Steel |