Learn More
asecos™ Stainless Steel Safety Canister
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
Brand: asecos™ 40091
Additional Details : Weight : 2.60000kg
Description
- Incl. multilingual safety sticker for content labelling, with filling tap made of stainless steel (1.4301)
- Incl. ventilation, NBR seals and aluminium screw cap integrated flame arrester and plastic protective ring
Specifications
2.6 gal. | |
Gray | |
9.61 in. | |
9.49 in. | |
5.73 lb. |
10 L | |
Stainless Steel | |
244.00 mm | |
241.00 mm | |
2.6 kg |