Learn More
Endecotts™ Stainless Steel Test Sieve, 100mm dia.
Full height test sieves provide all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieves, 100mm dia. comply with ISO 3310-1 / BS410 standards.
117.00 € - 179.00 €
Specifications
Diameter (Metric) Outer | 100 mm |
---|---|
Frame Material | Stainless steel |
Mesh Material | Stainless steel |
Certifications/Compliance | ISO 3310-1 / BS410 |
Height (Metric) | Full |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
12904225
|
Endecotts™
100SIW.038 |
38μm |
179.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12994215
|
Endecotts™
100SIW.045 |
45μm |
163.00 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
12974215
|
Endecotts™
100SIW.063 |
63μm |
129.00 €
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
12812753
|
Endecotts™
100SIW.075 |
75μm |
123.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12964215
|
Endecotts™
100SIW.090 |
90μm |
117.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12872743
|
Endecotts™
100SIW.106 |
106μm |
117.00 €
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
12822753
|
Endecotts™
100SIW.150 |
150μm |
117.00 €
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
12944215
|
Endecotts™
100SIW.180 |
180μm |
117.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
12882743
|
Endecotts™
100SIW.212 |
212μm |
127.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
12832753
|
Endecotts™
100SIW.300 |
300μm |
117.00 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
12892743
|
Endecotts™
100SIW.500 |
500μm |
117.00 €
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
12994205
|
Endecotts™
100SIW.710 |
710μm |
117.00 €
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
12984205
|
Endecotts™
100SIW.850 |
850μm |
117.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12964205
|
Endecotts™
100SIW1.18 |
1.18mm |
117.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12944205
|
Endecotts™
100SIW1.70 |
1.7mm |
117.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12934195
|
Endecotts™
100SIW11.2 |
11.2mm |
132.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
12924195
|
Endecotts™
100SIW13.2 |
13.2mm |
132.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12914195
|
Endecotts™
100SIW16.0 |
16mm |
132.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
12924205
|
Endecotts™
100SIW2.36 |
2.36mm |
117.00 €
Each |
Estimated Shipment: 23-04-2024
Log in to see stock. |
|||||
12904205
|
Endecotts™
100SIW3.35 |
3.35mm |
117.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12994195
|
Endecotts™
100SIW4.00 |
4mm |
132.00 €
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
12974195
|
Endecotts™
100SIW5.60 |
5.6mm |
132.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12964195
|
Endecotts™
100SIW6.70 |
6.7mm |
132.00 €
Each |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
12954195
|
Endecotts™
100SIW8.00 |
8mm |
132.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
Full height test sieves provide all types of laboratory sampling and particle size analysis. Endecotts™ Stainless Steel Test Sieves, 100mm dia. comply with ISO 3310-1 / BS410 standards.
- Stainless steel precision frame (ensures consistent nestability)
- Stainless steel mesh
- Precise aperture in accordance with ISO 3310-1 / BS410
- Natural fillet (free flowing of sample)
- Totally sealed (no crevice to lose material)
- Evenly tensioned mesh (ensures accurate analysis)
- Safe edge (big radius makes it comfortable to handle)
- Serial number (ensures full traceability)
Specifications
100 mm | |
Stainless steel | |
Full |
Stainless steel | |
ISO 3310-1 / BS410 |