Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50mmH, with Perforated Plates with Square Holes
Brand:  RETSCH 60.190.112000
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10032692
  • 215.00 € / Each
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Specifications
Stainless Steel
Stainless Steel
1 Each
Test Sieve
Certificate indicating Compliance
For separation, fractioning, particle size determination
200 mm
112mm
ISO 3310/2
50mm