Learn More
JP Selecta™ Stainless Steel Ultrasonic Cleaning Baths w/o Heating
Used for various applications like jewellery, watches, optical components, dentistry, and general laboratory use.JP Selecta™ Stainless Steel Ultrasonic Cleaning Baths has internal tank made from stainless steel.
POA
Description
Description
  • Internal tank made from AISI 304 stainless steel
  • Timer switch for 0 to 15 min, or continuous operation
  • Indicator lamp when unit is in operation