Learn More
Staurosporine, Fisher BioReagents
Brand: Fisher Bioreagents BP2541-500
Code : A0
Additional Details : CAS Number : 62996-74-1 Weight : 0.01000kg
Packaging | Poly Micro Tube |
---|---|
Quantity | 500μg |
Description
Staurosporine is a potent protein kinase inhibitor useful for studies on protein phosphorylation in the regulation of cellular functions.Chemical Identifiers
62996-74-1 | |
466.541 | |
staurosporine, kinome_3629 | |
CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC |
C28H26N4O3 | |
HKSZLNNOFSGOKW-ZYSRIHRCSA-N | |
49831000 |
Specifications
Staurosporine | |
Poly Micro Tube | |
White | |
500μg | |
62996-74-1 | |
C28H26N4O3 | |
staurosporine, kinome_3629 | |
CC12C(C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)NC)OC | |
49831000 | |
Solid |
Pass Test | |
Pass Test | |
270°C | |
From Streptomyces sp. | |
> 98.0 % | |
15, 8928 | |
HKSZLNNOFSGOKW-ZYSRIHRCSA-N | |
466.541 | |
466.5 | |
> 98.0% |
Safety and Handling
- Staurosporine
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
Hazard Statement
- H351-Suspected of causing cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.