missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Steel Bench Can

Brand:  Justrite™ 10291

Code : 03

Additional Details : Weight : 1.53600kg

 View more versions of this product

Product Code. 11953629

  • 91.88 € / Each


Estimated Shipment: 30-07-2021
Add to basket

Description

Description

Specifications

Specifications

Steel
4 L
267 mm
Videos
SDS
Documents
Certificates