missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Steel framed track support stand, width 600mm, T foot height 450/650
Brand: Manutan A341422
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Supports should be fitted every 1. 5m and at frame joints. Available as 'T' foot for bolting to floor, rubber foot, or castors with additional knee brace. Adjustable in height (add 100mm height for castors).- Track width: for 600mm track width
- Height: 450mm-650mm