Learn More
BD Biosciences™ Stem Cell Enumeration Kit (CE/IVD)
Provides simultaneous enumeration of viable dual-positive CD45+/CD34+ hematopoietic stem cell populations in CD34+ absolute counts (cells/μL) and the percentage of the total viable leucocyte count that is CD34+ (%CD34).
Brand: BD Biosciences™ 344563
Additional Details : Weight : 0.46000kg
Description
- Provides simultaneous enumeration of viable dual-positive CD45+/CD34+ hematopoietic stem cell populations in CD34+ absolute counts (cells/μL) as well as the percentage of the total viable leucocyte count that is CD34+ (%CD34).
- The following specimens can be analyzed with this kit: normal and mobilized peripheral blood, fresh and thawed leucopheresis products, fresh and thawed bone marrow, and fresh and thawed cord blood.
- The kit is intended for in vitro diagnostic (IVD) use on either a BD FACSCalibur™ flow cytometer using BD CellQuest™ or BD CellQuest™ Pro software or a BD FACSCanto™ II flow cytometer using BD FACSCanto™ software.
- Contents: BD Stem Cell reagent (CD45 FITC/CD34 PE), 7-aminoactinomycin-D (7-AAD) reagent, 10X ammonium chloride lysing solution, 50 BD Trucount tubes.
Specifications
50 Trucount Tubes | |
An unopened pouch is stable until the expiration date shown on the packaging. Do not open and use tubes after this expiration date. | |
Enumeration Kit |
BD Stem Cell reagent (CD45/CD34) provided in phosphate-buffered saline (PBS) containing bovine serum albumin (BSA) and 0.1% sodium azide, 7-AAD reagent, 10X ammonium chloride solution, 50 Trucount Tubes | |
50 Tests |