Learn More
Zeiss™ Stemi 305 Trinocular Stereo Microscope
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
2427.40 € - 4308.90 €
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17100686
|
Zeiss™
491903-0006-000 |
Double Spot Illuminator K LED |
2597.00 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
17322830
|
Zeiss™
491903-0007-000 |
Ring Illuminator K LED |
2427.40 €
1 set |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
17342830
|
Zeiss™
491903-0009-000 |
Ring Illuminator VisiLED |
4308.90 €
1 set |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Description
- This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
- The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
- Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Trinocular |