missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Step-on bin, yellow, 70 litre capacity, Ekwo
Brand: Manutan A021321
Code : Z3
Additional Details : Weight : 4.94000kg
Description
Lightweight yet highly robust plastic step-on bins enabling hygienic hands-free waste disposal. Rounded corners for easy cleaning. Supplied with two back wheels and a stabiliser at the front.- Capacity: 70L
- Compatible with 90 litre refuse bags
- Yellow
- Dimensions (H x W x D): 673mm x 495mm x 412mm