Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Brand:  Corning™ 6622
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15863895
  • 61.15 € / Pack of 100
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Specifications
Specifications
Autoclavable
Polypropylene
Dispensing of viscous and volatile liquids
100/Pk.
1.25 mL
ISO8655
Non-sterile
100
1.25 mL