Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Brand: Corning™ 6623
Additional Details : Weight : 5.00000kg
Descripción
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Especificaciones
Autoclavable | |
Polypropylene | |
Dispensing of viscous and volatile liquids | |
100/Pk. | |
2.5 mL |
ISO8655 | |
Non-sterile | |
100 | |
2.5 mL |