Learn More
Merck Stericup™ Quick Release Bottle Top Receiver Flasks
Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.
Brand:  Merck S200B10RE
Additional Details : Weight : 2.63300kg
 View more versions of this product
Product Code. 15987996
  • 166.00 € / Pack of 12
Estimated Shipment: 19-04-2024
to see stock.
Add to basket
Description
Description
Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.
Steritop™ bottle top filter units provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33 mm) or wide-mouth (45 mm) bottles.
A small tab inside the funnel secures an optional prefilter in-place during filtration.
For added protection against contamination, the threads for bottle attachment have been slightly recessed in case the funnel accidentally touches a work surface.
Specifications
Specifications
45 mm
1000 mL