missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Steriflip™ Sterile Disposable Vacuum Filter Units
Performance and convenience all in one. Steriflip™ Sterile Disposable Vacuum Filter Units uses vacuum to collect filtrate.
320.00 € - 324.00 €
Specifications
Color | Clear |
---|---|
Connectors | Double lead thread with vacuum port |
Disposable | Single-use |
Material | MBS/Polypropylene |
Sterility | Sterile |
Product Code | Brand | Material (Membrane) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Membrane) | Pore Size | Price | Quantity & Availability | ||||
10308522
|
Merck
SE1M179M6 |
PVDF | 0.22 μm |
320.00 €
Pack of 25 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
10541543
|
Merck
SE1M003M00 |
PVDF | 0.45 μm |
324.00 €
Pack of 25 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
Description
Performance and convenience all in one. Steriflip™ Sterile Disposable Vacuum Filter Units uses vacuum to collect filtrate.
OverviewThe Steriflip filter unit is designed to work with standard 50 mL centrifuge tubes, which eliminates sample transfer. To filter, just attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply the vacuum. When finished, the filtrate collects in the attached 50 mL centrifuge tube.
New pore sizes ideal for Cell Isolation applications
Steriflip Nylon Net filters are now available in 40, 60 and 100 μm pore sizes, making them ideal for stem cell, cardiomyocite, neurological and skeletal tissue applications. The vacuum-assisted, closed system option ensures sample sterility while enabling faster separation of large volumes of cellular material and improved recoveries.
The Steriflip™ filter unit eliminates messy pouring during sample transfer. Directly attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply vacuum. The filtrate collects in the attached 50 mL centrifuge tube. Steriflip™ filters come in different pore sizes that are ideal for cell isolation. These include nylon net filters in 40, 60 and 100 μm pore sizes, which are ideal for stem cell, cardiomyocyte, neurological and skeletal tissue applications. The vacuum-assisted, closed system ensures sample sterility while enabling fast separation of large volumes of cellular material for efficient recoveries.
Features & Benefits
- Eliminates messy sample transfer via a simple vacuum-assisted, closed system
- Isolate cells from cellular debris using any of the various pore-sized nylon net filters
- Ideal for sterilizing small batches of culture media
- Attaches to any standard 50 mL centrifuge tube, eliminating unnecesary materials, time and risk of spillage
Applications: Sterilization of Aqueous Solutions (up to 50 mL), Sterile Filtration of Small Batches of Culture Media, Sterile Filtration of Buffers with High Value Protein or Small Molecule Components, Cell Isolation
Order Info
Filter diameter: 40mm
Specifications
Clear | |
Single-use | |
Sterile | |
Low protein binding clarification of aqueous solutions | |
25 Pack | |
5.43 in. | |
4 cm |
Double lead thread with vacuum port | |
MBS/Polypropylene | |
7 cm2 | |
25Pack | |
50 mL | |
13.8 cm | |
6.4 cm |