Learn More
Technical Service Consultants™ Sterile Cotton Swabs
Use Technical Service Consultants™ Sterile Cotton Swabs to transport most fastidious and non-fastidious micro-organisms. They are also recommended for clinically significant samples.
Brand: Technical Service Consultants™ TS6-A
Code : 03
Additional Details : Weight : 0.20000kg
Description
- Extensive range of swabs available with a choice of swab shafts including white birchwood, polypropylene (flexible) and polystyrene (breakable)
- Tips formed from pure cotton, alginate and cotton
- Gamma irradiated to ensure a SAL of at least 10-6
Specifications
Wood | |
Sterile | |
In Tube | |
White | |
1 |
Cotton | |
2 Years | |
General swabbing & laboratory techniques. | |
500 | |
Swab |